siRNA Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142645.1 | NP_001136117.1 | 8 | 1079 | ||
| NR_136298.1 | 8 | 1033 | ||||
| XM_005246212.3 | XP_005246269.1 | 8 | 1233 | |||
| XM_005246213.3 | XP_005246270.1 | 8 | 918 | |||
| XM_011510458.2 | XP_011508760.1 | 8 | 1059 | |||
| XM_011510459.2 | XP_011508761.1 | 8 | 1264 | |||
| XM_017003098.1 | XP_016858587.1 | 8 | 1066 | |||
| XM_017003099.1 | XP_016858588.1 | 8 | 1066 | |||
| XM_017003100.1 | XP_016858589.1 | 8 | 1066 | |||
| GenBank mRNA | AK296987.1 | - | 8 | 1027 | ||

NEMP2
Genomic Map