siRNA Details
| Gene Symbol: SATB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125803.1 | 2 | 228 | |||
| GenBank mRNA | AW027730.1 | - | 2 | 192 | ||
| BF115425.1 | - | 2 | 228 | |||
| DR006704.1 | - | 2 | 447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SATB1-AS1| Gene Symbol: SATB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125803.1 | 2 | 228 | |||
| GenBank mRNA | AW027730.1 | - | 2 | 192 | ||
| BF115425.1 | - | 2 | 228 | |||
| DR006704.1 | - | 2 | 447 | |||