siRNA Details
| Gene Symbol: CTD-2151A2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110562.1 | 1 | 560 | |||
| GenBank mRNA | CR742340.1 | - | 1 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTD-2151A2.1| Gene Symbol: CTD-2151A2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110562.1 | 1 | 560 | |||
| GenBank mRNA | CR742340.1 | - | 1 | 560 | ||