siRNA Details
| Gene Symbol: C5orf63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164479.1 | NP_001157951.1 | 5 | 804 | ||
| GenBank mRNA | AK126569.1 | - | 6 | 902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5orf63| Gene Symbol: C5orf63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164479.1 | NP_001157951.1 | 5 | 804 | ||
| GenBank mRNA | AK126569.1 | - | 6 | 902 | ||