siRNA Details
| Gene Symbol: SMU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018225.2 | NP_060695.2 | 12 | 4963 | ||
| XM_005251503.4 | XP_005251560.1 | 11 | 4799 | |||
| GenBank mRNA | AL512690.1 | - | 1 | 821 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMU1| Gene Symbol: SMU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018225.2 | NP_060695.2 | 12 | 4963 | ||
| XM_005251503.4 | XP_005251560.1 | 11 | 4799 | |||
| GenBank mRNA | AL512690.1 | - | 1 | 821 | ||