siRNA Details
| Gene Symbol: EBLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036592.1 | 2 | 1372 | |||
| GenBank mRNA | AK023696.1 | - | 1 | 2417 | ||
| AK091718.1 | - | 2 | 1372 | |||
| BC032471.1 | - | 2 | 1271 | |||
| BC033624.1 | - | 2 | 1271 | |||
| BC080614.1 | - | 2 | 1250 | |||

EBLN3
Genomic Map