siRNA Details
| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024496.1 | 2 | 2862 | |||
| NR_135010.1 | 3 | 2281 | ||||
| GenBank mRNA | BC042539.1 | - | 2 | 2865 | ||
| BC068605.1 | - | 3 | 2283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTGER4P2-CDK2AP2P2| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024496.1 | 2 | 2862 | |||
| NR_135010.1 | 3 | 2281 | ||||
| GenBank mRNA | BC042539.1 | - | 2 | 2865 | ||
| BC068605.1 | - | 3 | 2283 | |||