siRNA Details
| Gene Symbol: SNX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012994.1 | NP_001013012.1 | 3 | 558 | ||
| XM_011518691.2 | XP_011516993.1 | 3 | 595 | |||
| XM_017014716.1 | XP_016870205.1 | 3 | 626 | |||
| GenBank mRNA | AK127013.1 | - | 2 | 262 | ||
| BX367137.2 | - | 3 | 557 | |||

SNX30
Genomic Map