siRNA Details
| Gene Symbol: MGC57346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026680.3 | 4 | 1560 | |||
| NR_027295.2 | 4 | 2054 | ||||
| GenBank mRNA | BC045575.1 | - | 4 | 2059 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC57346| Gene Symbol: MGC57346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026680.3 | 4 | 1560 | |||
| NR_027295.2 | 4 | 2054 | ||||
| GenBank mRNA | BC045575.1 | - | 4 | 2059 | ||