siRNA Details
Gene Symbol: MGC57346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026680.3 | 4 | 1386 | |||
NR_027295.2 | 4 | 1880 | ||||
GenBank mRNA | BC045575.1 | - | 4 | 1885 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MGC57346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026680.3 | 4 | 1386 | |||
NR_027295.2 | 4 | 1880 | ||||
GenBank mRNA | BC045575.1 | - | 4 | 1885 |