siRNA Details
| Gene Symbol: ZNF738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027130.1 | 5 | 7642 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF738| Gene Symbol: ZNF738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027130.1 | 5 | 7642 | |||