siRNA Details
| Gene Symbol: FBXW5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018998.3 | NP_061871.1 | 3, 4 | 528 | ||
| XM_017014812.1 | XP_016870301.1 | 3, 4 | 548 | |||
| GenBank mRNA | AK026081.1 | - | 3, 4 | 392 | ||
| AK315738.1 | - | 3, 4 | 403 | |||
| BC014130.1 | - | 3, 4 | 424 | 4620371 | ||
| BC014297.2 | - | 3, 4 | 395 | 3958395 | ||
| BC031100.1 | - | 3, 4 | 470 | |||
| BC035355.1 | - | 3, 4 | 462 | |||
| BI829307.1 | - | 3, 4 | 495 | |||
| CF132404.1 | - | 3, 4 | 469 | |||

FBXW5
Genomic Map