siRNA Details
| Gene Symbol: FER1L6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040044.1 | 6 | 1789 | |||
| GenBank mRNA | AK093040.1 | - | 6 | 1789 | ||
| BC137357.1 | - | 6 | 1756 | 9021355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FER1L6-AS1| Gene Symbol: FER1L6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040044.1 | 6 | 1789 | |||
| GenBank mRNA | AK093040.1 | - | 6 | 1789 | ||
| BC137357.1 | - | 6 | 1756 | 9021355 | ||