siRNA Details
| Gene Symbol: LINC00700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040253.2 | 2 | 232 | |||
| GenBank mRNA | AK097474.1 | - | 2 | 233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00700| Gene Symbol: LINC00700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040253.2 | 2 | 232 | |||
| GenBank mRNA | AK097474.1 | - | 2 | 233 | ||