siRNA Details
| Gene Symbol: HNRNPA1P33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003277.1 | 1 | 422 | |||
| GenBank mRNA | BC112994.1 | - | 1 | 422 | ||
| BC112995.1 | - | 1 | 422 | |||
| BC128038.1 | - | 1 | 422 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HNRNPA1P33| Gene Symbol: HNRNPA1P33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003277.1 | 1 | 422 | |||
| GenBank mRNA | BC112994.1 | - | 1 | 422 | ||
| BC112995.1 | - | 1 | 422 | |||
| BC128038.1 | - | 1 | 422 | |||