siRNA Details
| Gene Symbol: KAZALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319303.1 | NP_001306232.1 | 6 | 2712 | ||
| NR_135067.1 | 5 | 2095 | ||||
| NR_135068.1 | 2 | 1044 | ||||
| XM_017016714.1 | XP_016872203.1 | 5 | 1736 | |||
| XM_017016715.1 | XP_016872204.1 | 5 | 1713 | |||
| GenBank mRNA | AF333487.1 | - | 3 | 837 | ||
| AK172864.1 | - | 7 | 2623 | |||
| AY014271.1 | - | 3 | 837 | |||
| BC012394.1 | - | 2 | 944 | |||

KAZALD1
Genomic Map