siRNA Details
| Gene Symbol: FZD10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033834.1 | 4 | 2589 | |||
| GenBank mRNA | AK056047.1 | - | 4 | 1844 | ||
| AK056281.1 | - | 2 | 1048 | |||
| AK123838.1 | - | 4 | 2589 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FZD10-AS1| Gene Symbol: FZD10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033834.1 | 4 | 2589 | |||
| GenBank mRNA | AK056047.1 | - | 4 | 1844 | ||
| AK056281.1 | - | 2 | 1048 | |||
| AK123838.1 | - | 4 | 2589 | |||