siRNA Details
Gene Symbol: SMIM10L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271592.1 | NP_001258521.1 | 1 | 807 | ||
GenBank mRNA | BX338800.2 | - | 1 | 773 | ||
CX164259.1 | - | 1 | 741 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMIM10L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271592.1 | NP_001258521.1 | 1 | 807 | ||
GenBank mRNA | BX338800.2 | - | 1 | 773 | ||
CX164259.1 | - | 1 | 741 |