siRNA Details
| Gene Symbol: ZSCAN29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152455.3 | NP_689668.3 | 1 | 53 | ||
| XM_006720401.3 | XP_006720464.1 | 2 | 813 | |||
| XM_011521266.2 | XP_011519568.1 | 2 | 493 | |||
| XM_017021949.1 | XP_016877438.1 | 1 | 1025 | |||
| GenBank mRNA | AK122788.1 | - | 1 | 41 | ||
| BC043612.1 | - | 1 | 55 | |||

ZSCAN29
Genomic Map