siRNA Details
| Gene Symbol: LOC105371184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136337.1 | 4 | 700 | |||
| GenBank mRNA | BC004968.1 | - | 1 | 1204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371184| Gene Symbol: LOC105371184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136337.1 | 4 | 700 | |||
| GenBank mRNA | BC004968.1 | - | 1 | 1204 | ||