siRNA Details
| Gene Symbol: CCDC93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019044.4 | NP_061917.3 | 4 | 463 | ||
| XM_006712600.2 | XP_006712663.1 | 4 | 463 | |||
| XM_011511359.1 | XP_011509661.1 | 4 | 463 | |||
| XM_011511361.1 | XP_011509663.1 | 3 | 217 | |||
| XM_011511363.2 | XP_011509665.1 | 4 | 463 | |||
| GenBank mRNA | AB209993.1 | - | 4 | 382 | ||
| AK025611.1 | - | 3 | 231 | |||
| AK057926.1 | - | 4 | 324 | |||
| AK290722.1 | - | 4 | 370 | |||
| AK309040.1 | - | 4 | 370 | |||
| BC028609.1 | - | 4 | 436 | 4826022 | ||
| BC093018.1 | - | 4 | 353 | 30337330 | ||
| DB084596.1 | - | 4 | 463 | |||

CCDC93
Genomic Map