siRNA Details
| Gene Symbol: CDIPT-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015396.1 | 5, 6 | 566 | |||
| NR_024370.1 | 5, 6 | 443 | ||||
| GenBank mRNA | AK131031.1 | - | 5, 6 | 566 | ||
| BC070380.1 | - | 5, 6 | 548 | |||
| BM006535.1 | - | 5, 6 | 444 | |||

CDIPT-AS1
Genomic Map