siRNA Details
| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105079.2 | NP_001098549.2 | 2, 3 | 632 | ||
| XM_011545916.2 | XP_011544218.1 | 2, 3 | 2009 | |||
| XM_011545917.2 | XP_011544219.1 | 2, 3 | 2009 | |||
| GenBank mRNA | DB082271.1 | - | 2, 3 | 229 | ||

FBRS
Genomic Map