siRNA Details
| Gene Symbol: FAM104B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166700.1 | NP_001160172.1 | 3 | 399 | ||
| NM_001166701.1 | NP_001160173.1 | 3 | 396 | |||
| NM_001166702.1 | NP_001160174.1 | 3 | 497 | |||
| NM_001166703.1 | NP_001160175.1 | 3 | 575 | |||
| NM_001166704.1 | NP_001160176.1 | 3 | 403 | |||
| XM_017029951.1 | XP_016885440.1 | 3 | 420 | |||
| XM_017029952.1 | XP_016885441.1 | 3 | 633 | |||
| GenBank mRNA | AK299935.1 | - | 3 | 484 | ||
| AK311333.1 | - | 2 | 285 | |||
| AK311699.1 | - | 3 | 390 | |||
| BC006406.2 | - | 1 | 358 | |||
| BX100076.1 | - | 3 | 575 | |||
| DB216586.1 | - | 3 | 396 | |||
| DC413808.1 | - | 3 | 497 | |||

FAM104B
Genomic Map