siRNA Details
| Gene Symbol: FAM104B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166700.1 | NP_001160172.1 | 3 | 349 | ||
| NM_001166701.1 | NP_001160173.1 | 3 | 346 | |||
| NM_001166702.1 | NP_001160174.1 | 3 | 447 | |||
| NM_001166703.1 | NP_001160175.1 | 3 | 525 | |||
| NM_001166704.1 | NP_001160176.1 | 3 | 353 | |||
| XM_017029951.1 | XP_016885440.1 | 3 | 370 | |||
| XM_017029952.1 | XP_016885441.1 | 3 | 583 | |||
| GenBank mRNA | AK299935.1 | - | 3 | 434 | ||
| AK311333.1 | - | 2 | 235 | |||
| AK311699.1 | - | 3 | 340 | |||
| BC006406.2 | - | 1 | 308 | |||
| BX100076.1 | - | 3 | 525 | |||
| DB216586.1 | - | 3 | 346 | |||
| DC413808.1 | - | 3 | 447 | |||

FAM104B
Genomic Map