siRNA Details
| Gene Symbol: CNIH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322304.1 | NP_001309233.1 | 2 | 385 | ||
| NR_136289.1 | 2 | 315 | ||||
| NR_136290.1 | 2 | 315 | ||||
| NR_136291.1 | 3 | 451 | ||||
| NR_136292.1 | 2 | 385 | ||||
| NR_136293.1 | 4 | 555 | ||||
| NR_136294.1 | 2 | 385 | ||||
| NR_136296.1 | 2 | 385 | ||||

CNIH3
Genomic Map