siRNA Details
| Gene Symbol: FAM212B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019099.4 | NP_061972.1 | 2 | 259 | ||
| NM_198926.2 | NP_945120.1 | 2 | 117 | |||
| XM_011541783.2 | XP_011540085.1 | 4 | 355 | |||
| GenBank mRNA | AK055667.1 | - | 2 | 235 | ||
| AK293989.1 | - | 2 | 144 | |||
| AK295012.1 | - | 2 | 198 | |||
| AL137198.1 | - | 2 | 166 | |||
| AL137199.1 | - | 2 | 117 | |||
| BC031558.1 | - | 2 | 214 | 5181834 | ||
| DA306915.1 | - | 2 | 259 | |||

FAM212B
Genomic Map