siRNA Details
| Gene Symbol: LOC107985555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017029168.1 | XP_016884657.1 | 4 | 912 | ||
| XM_017029169.1 | XP_016884658.1 | 3 | 1041 | |||
| XM_017029170.1 | XP_016884659.1 | 4 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107985555| Gene Symbol: LOC107985555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017029168.1 | XP_016884657.1 | 4 | 912 | ||
| XM_017029169.1 | XP_016884658.1 | 3 | 1041 | |||
| XM_017029170.1 | XP_016884659.1 | 4 | 477 | |||