siRNA Details
| Gene Symbol: MIAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003491.3 | 3 | 1010 | |||
| NR_033319.2 | 3 | 1010 | ||||
| NR_033320.2 | 3 | 936 | ||||
| NR_033321.2 | 3 | 936 | ||||
| GenBank mRNA | AB263414.1 | - | 3 | 972 | ||
| AB263415.1 | - | 3 | 972 | |||
| AB263416.1 | - | 3 | 898 | |||
| AB263417.1 | - | 3 | 898 | |||
| AK098833.1 | - | 3 | 904 | |||

MIAT
Genomic Map