siRNA Details
| Gene Symbol: SHISA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207020.1 | NP_001193949.1 | 4 | 1088 | ||
| XM_006724254.3 | XP_006724317.1 | 4 | 1799 | |||
| XM_006724255.2 | XP_006724318.1 | 4 | 1691 | |||
| XM_006724256.3 | XP_006724319.1 | 4 | 1336 | |||

SHISA8
Genomic Map