siRNA Details
Gene Symbol: LOC101926966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110201.1 | 4 | 458 | |||
GenBank mRNA | AK093525.1 | - | 4 | 458 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101926966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110201.1 | 4 | 458 | |||
GenBank mRNA | AK093525.1 | - | 4 | 458 |