siRNA Details
| Gene Symbol: LOC101926966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110201.1 | 4 | 456 | |||
| GenBank mRNA | AK093525.1 | - | 4 | 456 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926966| Gene Symbol: LOC101926966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110201.1 | 4 | 456 | |||
| GenBank mRNA | AK093525.1 | - | 4 | 456 | ||