siRNA Details
| Gene Symbol: IQCF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143833.3 | NP_001137305.2 | 1 | 130 | ||
| XM_005265168.3 | XP_005265225.1 | 2 | 697 | |||
| XM_011533728.1 | XP_011532030.1 | 2 | 168 | |||
| GenBank mRNA | BX093815.1 | - | 1 | 130 | ||

IQCF6
Genomic Map