siRNA Details
| Gene Symbol: ROBO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128929.3 | NP_001122401.1 | 2 | 369 | ||
| XM_017006990.1 | XP_016862479.1 | 2 | 369 | |||
| XM_017006992.1 | XP_016862481.1 | 2 | 369 | |||
| XM_017006996.1 | XP_016862485.1 | 2 | 369 | |||
| XM_017006997.1 | XP_016862486.1 | 2 | 369 | |||
| XM_017007004.1 | XP_016862493.1 | 2 | 369 | |||
| XM_017007005.1 | XP_016862494.1 | 2 | 369 | |||
| GenBank mRNA | AK301176.1 | - | 2 | 242 | ||
| DQ533873.1 | - | 2 | 350 | |||

ROBO2
Genomic Map