siRNA Details
Gene Symbol: LINC00971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033860.1 | 21 | 6905 | |||
GenBank mRNA | BC068246.1 | - | 21 | 6907 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033860.1 | 21 | 6905 | |||
GenBank mRNA | BC068246.1 | - | 21 | 6907 |