siRNA Details
| Gene Symbol: LINC00971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033860.1 | 21 | 7031 | |||
| GenBank mRNA | BC068246.1 | - | 21 | 7033 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00971| Gene Symbol: LINC00971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033860.1 | 21 | 7031 | |||
| GenBank mRNA | BC068246.1 | - | 21 | 7033 | ||