siRNA Details
| Gene Symbol: COL6A4P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027898.1 | 12, 13 | 665 | |||
| GenBank mRNA | CN285077.1 | - | 10, 11 | 569 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

COL6A4P2| Gene Symbol: COL6A4P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027898.1 | 12, 13 | 665 | |||
| GenBank mRNA | CN285077.1 | - | 10, 11 | 569 | ||