siRNA Details
| Gene Symbol: ETAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019002.3 | NP_061875.2 | 5 | 788 | ||
| XM_017004376.1 | XP_016859865.1 | 5 | 781 | |||
| XM_017004377.1 | XP_016859866.1 | 5 | 781 | |||
| GenBank mRNA | AJ242682.2 | - | 5 | 793 | ||
| AK025155.1 | - | 4 | 321 | |||
| BC033075.1 | - | 5 | 760 | |||
| BC040001.1 | - | 5 | 720 | 5556133 | ||

ETAA1
Genomic Map