siRNA Details
| Gene Symbol: LINC00613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103762.1 | 3 | 275 | |||
| NR_103763.1 | 2 | 115 | ||||
| GenBank mRNA | AW237894.1 | - | 3 | 296 | ||
| BG204498.1 | - | 1 | 46 | |||
| BG220241.1 | - | 2 | 115 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00613| Gene Symbol: LINC00613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103762.1 | 3 | 275 | |||
| NR_103763.1 | 2 | 115 | ||||
| GenBank mRNA | AW237894.1 | - | 3 | 296 | ||
| BG204498.1 | - | 1 | 46 | |||
| BG220241.1 | - | 2 | 115 | |||