siRNA Details
| Gene Symbol: FLJ31104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102755.1 | 2 | 1417 | |||
| GenBank mRNA | AK055666.1 | - | 2 | 1264 | ||
| BC039324.1 | - | 2 | 1417 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ31104| Gene Symbol: FLJ31104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102755.1 | 2 | 1417 | |||
| GenBank mRNA | AK055666.1 | - | 2 | 1264 | ||
| BC039324.1 | - | 2 | 1417 | |||