siRNA Details
| Gene Symbol: NR2F1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124699.1 | - | 1 | 560 | ||
| BC144330.1 | - | 1 | 1110 | |||
| BC144331.1 | - | 1 | 1110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR2F1-AS1| Gene Symbol: NR2F1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124699.1 | - | 1 | 560 | ||
| BC144330.1 | - | 1 | 1110 | |||
| BC144331.1 | - | 1 | 1110 | |||