siRNA Details
| Gene Symbol: CCDC192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317938.1 | NP_001304867.1 | 6 | 528 | ||
| XM_017009805.1 | XP_016865294.1 | 5 | 590 | |||
| XM_017009806.1 | XP_016865295.1 | 6 | 494 | |||
| XM_017009807.1 | XP_016865296.1 | 6 | 485 | |||
| XM_017009808.1 | XP_016865297.1 | 5 | 416 | |||
| XM_017009809.1 | XP_016865298.1 | 6 | 559 | |||
| XM_017009810.1 | XP_016865299.1 | 6 | 558 | |||
| XM_017009811.1 | XP_016865300.1 | 6 | 558 | |||

CCDC192
Genomic Map