siRNA Details
| Gene Symbol: FAM64A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195228.1 | NP_001182157.1 | 5 | 829 | ||
| NM_019013.2 | NP_061886.2 | 6 | 874 | |||
| XM_017024778.1 | XP_016880267.1 | 4 | 1203 | |||
| GenBank mRNA | AK001018.1 | - | 5 | 814 | ||
| AK001353.1 | - | 5 | 789 | |||
| AK225613.1 | - | 6 | 836 | |||
| AK225702.1 | - | 6 | 834 | |||
| BC005004.1 | - | 5 | 796 | 3533875 | ||
| BC013966.2 | - | 6 | 820 | 2988318 | ||
| KU178653.1 | - | 4 | 720 | |||
| KU178654.1 | - | 5 | 765 | |||

FAM64A
Genomic Map