siRNA Details
| Gene Symbol: FAM105A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019018.2 | NP_061891.1 | 2 | 257 | ||
| XM_011514056.2 | XP_011512358.1 | 2 | 239 | |||
| GenBank mRNA | AK001989.1 | - | 2 | 216 | ||
| AK222731.1 | - | 2 | 244 | |||
| BC011524.1 | - | 2 | 183 | 3872837 | ||

FAM105A
Genomic Map