siRNA Details
| Gene Symbol: ERICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534735.2 | XP_011533037.1 | 6 | 2532 | ||
| XM_017013127.1 | XP_016868616.1 | 8 | 1755 | |||
| XM_017013132.1 | XP_016868621.1 | 7 | 1557 | |||
| GenBank mRNA | AK090404.1 | - | 3 | 4697 | ||
| AK125470.1 | - | 3 | 1444 | |||

ERICH1
Genomic Map