siRNA Details
| Gene Symbol: LOC101927752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI590306.1 | - | 1, 2 | 298 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927752| Gene Symbol: LOC101927752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI590306.1 | - | 1, 2 | 298 | ||