siRNA Details
| Gene Symbol: ELP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288725.1 | NP_001275654.1 | 3 | 380 | ||
| NM_001288726.1 | NP_001275655.1 | 3 | 380 | |||
| NM_019040.4 | NP_061913.3 | 3 | 380 | |||
| GenBank mRNA | AK000505.1 | - | 3 | 359 | ||
| AK304885.1 | - | 3 | 339 | |||
| AL136677.1 | - | 3 | 337 | |||
| BC012514.1 | - | 3 | 331 | 4509903 | ||
| HY127287.1 | - | 3 | 382 | |||

ELP4
Genomic Map