siRNA Details
Gene Symbol: LINC00268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC136786.1 | - | 2 | 1998 | 9020776 | |
BC136790.1 | - | 2 | 1999 | 9020780 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC136786.1 | - | 2 | 1998 | 9020776 | |
BC136790.1 | - | 2 | 1999 | 9020780 |