siRNA Details
| Gene Symbol: LINC00891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034005.1 | 3 | 467 | |||
| GenBank mRNA | AK128305.1 | - | 3 | 467 | ||
| Gene Symbol: LOC100132741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034004.1 | 3 | 467 | |||

LINC00891
Genomic Map