siRNA Details
Gene Symbol: LINC00684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034116.1 | 1 | 107 | |||
NR_120499.1 | 1 | 107 | ||||
GenBank mRNA | AA393776.1 | - | 1 | 63 | ||
HY038238.1 | - | 1 | 65 |
Gene Symbol: LOC100132304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120493.1 | 1 | 107 | |||
NR_120494.1 | 1 | 107 | ||||
GenBank mRNA | AA436071.1 | - | 1 | 44 | ||
HY042459.1 | - | 1 | 65 |
